CAS RN 32841-83-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,1'-Ferrocenediboronic Acid (contains varying amounts of Anhydride) 95.0+%, TCI America™
CAS: 32841-83-1 Molecular Formula: C10H12B2FeO4