accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32852-81-6

IUPAC Name: 2-(3-phenoxyphenyl)acetic acid
Synonyms: (3-phenoxyphenyl)acetic acid
Molecular Weight (g/mol): 228.25
Molecular Formula: C14H12O3
InChi Key: LEMRHTTWKDVQEI-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC(OC2=CC=CC=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results