accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOFFF

CAS RN 32857-62-8

IUPAC Name: 2-[4-(trifluoromethyl)phenyl]acetic acid
Synonyms: [4-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: HNORVZDAANCHAY-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(C=C1)C(F)(F)F
Percent Purity 
  • (3)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)