accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32860-56-3

IUPAC Name: [31,32,33,34,35,36,37,38,39,40,41,42-dodecahydroxy-10,15,20,25,30-pentakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontan-5-yl]methyl 4-methylbenzene-1-sulfonate
Synonyms: [31,32,33,34,35,36,37,38,39,40,41,42-dodecahydroxy-10,15,20,25,30-pentakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontan-5-yl]methyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 1127.03
Molecular Formula: C43H66O32S
InChi Key: ARQITQMHQNGIEE-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC1C(O)C2O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)