accessibility menu, dialog, popup

UserName

Viewing profile as user:
NONO

CAS RN 32863-33-5

IUPAC Name: 2,1,3-benzoxadiazole-5-carbaldehyde
Synonyms: 2,1,3-benzoxadiazole-5-carbaldehyde
Molecular Weight (g/mol): 148.12
Molecular Formula: C7H4N2O2
InChi Key: STVDCFOBQWMSHN-UHFFFAOYSA-N
SMILES: O=CC1=CC2=NON=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results