accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32887-01-7

IUPAC Name: (2S,5R,6R)-6-{[(azepan-1-yl)methylidene]amino}-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Synonyms: mecillinam coactin mecilinamo penicillin HX mecillinamum
Molecular Weight (g/mol): 325.43
Molecular Formula: C15H23N3O3S
InChi Key: BWWVAEOLVKTZFQ-NTZNESFSSA-N
SMILES: CC1(C)S[C@@H]2[C@H](N=CN3CCCCCC3)C(=O)N2[C@H]1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results