accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32894-07-8

IUPAC Name: 1-(benzylsulfanyl)ethan-1-imine hydrochloride
Synonyms: 1-(benzylsulfanyl)ethanimine hydrochloride
Molecular Weight (g/mol): 201.71
Molecular Formula: C9H12ClNS
InChi Key: OVQWHRFBVPDPCD-UHFFFAOYSA-N
SMILES: Cl.CC(=N)SCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results