accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3H2COOH3CCH2OOCH3CH2

CAS RN 3290-92-4

IUPAC Name: 2,2-bis({[(2-methylprop-2-enoyl)oxy]methyl})butyl 2-methylprop-2-enoate
Synonyms: TMPT light ester tmp tmpt (crosslinking agent) acryester tmp chemlink 30 sartomer 350 NK ester tmpt saret 515 PTMA tmptma
Molecular Weight (g/mol): 338.4
Molecular Formula: C18H26O6
InChi Key: OKKRPWIIYQTPQF-UHFFFAOYSA-N
SMILES: CCC(COC(=O)C(C)=C)(COC(=O)C(C)=C)COC(=O)C(C)=C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results