accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32916-51-1

IUPAC Name: chloro(cyclopentyl)magnesium
Synonyms: chloro(cyclopentyl)magnesium
Molecular Weight (g/mol): 128.88
Molecular Formula: C5H9ClMg
InChi Key: GROGEABZCKQTJW-UHFFFAOYSA-M
SMILES: Cl[Mg]C1CCCC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)