accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNOBr

CAS RN 3292-77-1

IUPAC Name: 2-bromo-1-(1,3-thiazol-2-yl)ethan-1-one
Synonyms: 2-bromo-1-(1,3-thiazol-2-yl)ethanone
Molecular Weight (g/mol): 206.06
Molecular Formula: C5H4BrNOS
InChi Key: AQRFTRDAOYSMEA-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results