accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHNNOHO

CAS RN 32926-43-5

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-[1-(triphenylmethyl)-1H-imidazol-4-yl]propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-[1-(triphenylmethyl)imidazol-4-yl]propanoic acid
Molecular Weight (g/mol): 497.6
Molecular Formula: C30H31N3O4
InChi Key: OYXZPXVCRAAKCM-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results