accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3294-03-9

IUPAC Name: 2-{[2-hydroxy-5-(2,4,4-trimethylpentan-2-yl)phenyl]sulfanyl}-4-(2,4,4-trimethylpentan-2-yl)phenol
Synonyms: 2-{[2-hydroxy-5-(2,4,4-trimethylpentan-2-yl)phenyl]sulfanyl}-4-(2,4,4-trimethylpentan-2-yl)phenol
Molecular Weight (g/mol): 442.7
Molecular Formula: C28H42O2S
InChi Key: WQYFETFRIRDUPJ-UHFFFAOYSA-N
SMILES: CC(C)(C)CC(C)(C)C1=CC=C(O)C(SC2=CC(=CC=C2O)C(C)(C)CC(C)(C)C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results