accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 32940-15-1

IUPAC Name: 5-methoxy-1,2,3,4-tetrahydronaphthalen-2-one
Synonyms: 5-methoxy-3,4-dihydro-1H-naphthalen-2-one
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: MDAIAXRTLTVEOU-UHFFFAOYSA-N
SMILES: COC1=CC=CC2=C1CCC(=O)C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results