accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONN

CAS RN 32959-62-9

IUPAC Name: 2-(pyridin-2-yl)-1,3-benzoxazole
Synonyms: 2-(pyridin-2-yl)-1,3-benzoxazole
Molecular Weight (g/mol): 196.21
Molecular Formula: C12H8N2O
InChi Key: WELSCYIRWKBEBZ-UHFFFAOYSA-N
SMILES: O1C2=CC=CC=C2N=C1C1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results