accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 329689-23-8

IUPAC Name: ethyl 4-(3-hydroxyphenyl)-6-methyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidine-5-carboxylate
Synonyms: monastrol
Molecular Weight (g/mol): 292.35
Molecular Formula: C14H16N2O3S
InChi Key: LOBCDGHHHHGHFA-UHFFFAOYNA-N
SMILES: CCOC(=O)C1=C(C)NC(=S)NC1C1=CC(O)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results