accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(S)O(R)(S)OH(S)CH3(R)(S)OO(S)OH(S)HO(R)HOOCH3CH3H3C

CAS RN 32981-86-5

IUPAC Name: (1S,2S,3R,4S,7R,9S,10S,12R,15S)-4-(acetyloxy)-1,9,12,15-tetrahydroxy-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate
Synonyms: 10-deacetylbaccatin III
Molecular Weight (g/mol): 544.6
Molecular Formula: C29H36O10
InChi Key: YWLXLRUDGLRYDR-ZHPRIASZSA-N
SMILES: CC(=O)O[C@@]12CO[C@@H]1C[C@H](O)[C@]1(C)[C@@H]2[C@H](OC(=O)C2=CC=CC=C2)[C@]2(O)C[C@H](O)C(C)=C([C@@H](O)C1=O)C2(C)C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)