accessibility menu, dialog, popup

UserName

Viewing profile as user:
RuClPPOCH3PhPhPhPhPhPh

CAS RN 32993-05-8

IUPAC Name: Chloro(cyclopentadienyl)bis(triphenylphosphine)ruthenium(II) ethanol adduct
Synonyms:
Molecular Weight (g/mol): 772.27
Molecular Formula: C43H41ClOP2Ru
InChi Key: YKZUFRITHQLOHK-UHFFFAOYSA-M
SMILES: Cl[Ru++].CCO.c1cccc1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (3)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)