accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 329940-78-5

IUPAC Name: 2-chloro-N-(2-methylphenyl)-4-nitrobenzamide
Synonyms: 2-chloro-N-(2-methylphenyl)-4-nitrobenzamide
Molecular Weight (g/mol): 290.7
Molecular Formula: C14H11ClN2O3
InChi Key: XQBCDIDTLUWUMG-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1NC(=O)C1=C(Cl)C=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results