accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 330-84-7

IUPAC Name: 1-fluoro-4-phenoxybenzene
Synonyms: 1-fluoro-4-phenoxybenzene
Molecular Weight (g/mol): 188.2
Molecular Formula: C12H9FO
InChi Key: AODSTUBSNYVSSL-UHFFFAOYSA-N
SMILES: FC1=CC=C(OC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results