CAS RN 33006-91-6
IUPAC Name:
4-[2-(5-chloropyridin-2-yl)diazen-1-yl]benzene-1,3-diamine
Synonyms:
4-[2-(5-chloropyridin-2-yl)diazen-1-yl]benzene-1,3-diamine
Molecular Weight (g/mol):
247.69
Molecular Formula:
C11H10ClN5
InChi Key:
WLNTVTDTBKPTCA-UHFFFAOYSA-N
SMILES:
NC1=CC=C(N=NC2=CC=C(Cl)C=N2)C(N)=C1
1
–
1
of
1
results
1
4-(5-Chloro-2-pyridylazo)-1,3-phenylenediamine 99.0+%, TCI America™
CAS: 33006-91-6 Molecular Formula: C11H10ClN5 Molecular Weight (g/mol): 247.686 MDL Number: MFCD00059781 InChI Key: WLNTVTDTBKPTCA-UHFFFAOYSA-N PubChem CID: 118377 IUPAC Name: 4-[(5-chloropyridin-2-yl)diazenyl]benzene-1,3-diamine SMILES: C1=CC(=C(C=C1N)N)N=NC2=NC=C(C=C2)Cl