accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33016-47-6

IUPAC Name: 1-(triphenylmethyl)-1H-imidazole-4-carbaldehyde
Synonyms: 1-(triphenylmethyl)imidazole-4-carbaldehyde
Molecular Weight (g/mol): 338.41
Molecular Formula: C23H18N2O
InChi Key: YQYLLBSWWRWWAY-UHFFFAOYSA-N
SMILES: O=CC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)