accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33018-91-6

IUPAC Name: 7-ethoxy-7-oxoheptanoic acid
Synonyms: heptanedioic acid monoethyl ester
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: NQYXFXWKKYGBNL-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCCCC(O)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)