accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COPNPNPNFFFFF

CAS RN 33027-66-6

IUPAC Name: 2-ethoxy-2,4,4,6,6-pentafluoro-1,3,5,2λ⁵,4λ⁵,6λ⁵-triazatriphosphinine
Synonyms: 2-ethoxy-2,4,4,6,6-pentafluoro-1,3,5,2λ⁵,4λ⁵,6λ⁵-triazatriphosphinine
Molecular Weight (g/mol): 275
Molecular Formula: C2H5F5N3OP3
InChi Key: CBTAIOOTRCAMBD-UHFFFAOYSA-N
SMILES: CCOP1(F)=NP(F)(F)=NP(F)(F)=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results