accessibility menu, dialog, popup

UserName

Viewing profile as user:
OPNPNPNFFFFF

CAS RN 33027-68-8

IUPAC Name: 2,2,4,4,6-pentafluoro-6-phenoxy-1,3,5,2λ⁵,4λ⁵,6λ⁵-triazatriphosphinine
Synonyms: 2,2,4,4,6-pentafluoro-6-phenoxy-1,3,5,2λ⁵,4λ⁵,6λ⁵-triazatriphosphinine
Molecular Weight (g/mol): 323.04
Molecular Formula: C6H5F5N3OP3
InChi Key: XNZZEQCBAGUFMT-UHFFFAOYSA-N
SMILES: FP1(F)=NP(F)(OC2=CC=CC=C2)=NP(F)(F)=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results