accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CH2NOHNOHOHO

CAS RN 3303-29-5

IUPAC Name: 2-(2-amino-4-methylpentanamido)-3-(4-hydroxyphenyl)propanoic acid
Synonyms: leu-tyr
Molecular Weight (g/mol): 294.35
Molecular Formula: C15H22N2O4
InChi Key: LHSGPCFBGJHPCY-UHFFFAOYNA-N
SMILES: CC(C)CC(N)C(=O)NC(CC1=CC=C(O)C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results