accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOCH3SNH2OO

CAS RN 33045-52-2

IUPAC Name: methyl 2-methoxy-5-sulfamoylbenzoate
Synonyms: methyl 2-methoxy-5-sulfamoylbenzoate
Molecular Weight (g/mol): 245.25
Molecular Formula: C9H11NO5S
InChi Key: MKDYDRQLKPGNNU-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC=C1OC)S(N)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results