accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SH2NOO

CAS RN 3306-62-5

IUPAC Name: 2-aminobenzene-1-sulfonamide
Synonyms: 2-aminobenzenesulfonamide BENZENESULFONAMIDE2amino
Molecular Weight (g/mol): 172.2
Molecular Formula: C6H8N2O2S
InChi Key: YAZSBRQTAHVVGE-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1S(N)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)