accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOHOOPOOOOHHOHO

CAS RN 33068-18-7

IUPAC Name: disodium (3,4,5,6-tetrahydroxyoxan-2-yl)methyl phosphate
Synonyms: disodium (3,4,5,6-tetrahydroxyoxan-2-yl)methyl phosphate
Molecular Weight (g/mol): 304.1
Molecular Formula: C6H11Na2O9P
InChi Key: CHIBNKGHYAQTQY-UHFFFAOYNA-L
SMILES: [Na+].[Na+].OC1OC(COP([O-])([O-])=O)C(O)C(O)C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results