accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFOBrO

CAS RN 33070-32-5

IUPAC Name: 5-bromo-2,2-difluoro-2H-1,3-benzodioxole
Synonyms: 5-bromo-2,2-difluoro-1,3-benzodioxole
Molecular Weight (g/mol): 237
Molecular Formula: C7H3BrF2O2
InChi Key: SZRHWHHXVXSGMT-UHFFFAOYSA-N
SMILES: FC1(F)OC2=CC=C(Br)C=C2O1
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)