accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3OOHNBOH3CH3CH3CCH3O

CAS RN 330793-01-6

IUPAC Name: tert-butyl N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
Synonyms: tert-butyl N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
Molecular Weight (g/mol): 319.21
Molecular Formula: C17H26BNO4
InChi Key: HSJNIOYPTSKQBD-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (6)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)