CAS RN 330794-03-1
IUPAC Name:
4-bromo-5-fluoro-2-methoxyaniline
Synonyms:
4-bromo-5-fluoro-2-methoxyaniline
Molecular Weight (g/mol):
220.04
Molecular Formula:
C7H7BrFNO
InChi Key:
WOFOKKKWMDHLEX-UHFFFAOYSA-N
SMILES:
COC1=CC(Br)=C(F)C=C1N
1
–
1
of
1
results
1
Promotions Available
4-Bromo-5-fluoro-2-methoxyaniline, 96%, Thermo Scientific Chemicals
4-Bromo-5-fluoro-2-methoxyaniline, 96%, Thermo Scientific Chemicals
CAS: 330794-03-1 Molecular Formula: C7H7BrFNO Molecular Weight (g/mol): 220.04 MDL Number: MFCD21603958 InChI Key: WOFOKKKWMDHLEX-UHFFFAOYSA-N Synonym: benzenamine, 4-bromo-5-fluoro-2-methoxy PubChem CID: 22347298 IUPAC Name: 4-bromo-5-fluoro-2-methoxyaniline SMILES: COC1=CC(Br)=C(F)C=C1N