accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COONHBOH3CH3CH3CCH3O

CAS RN 330794-35-9

IUPAC Name: tert-butyl N-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}carbamate
Synonyms: tert-butyl N-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}carbamate
Molecular Weight (g/mol): 333.24
Molecular Formula: C18H28BNO4
InChi Key: CUDCEJRRWNIPDL-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results