CAS RN 330796-24-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Hedgehog Antagonist VIII
The Hedgehog Antagonist VIII, also referenced under CAS 330796-24-2, controls the biological activity of Hedgehog. This small molecule/inhibitor is primarily used for Cell Signaling applications.