accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNCH3H3CNCH3CH3(E/Z)(E/Z)

CAS RN 33089-61-1

IUPAC Name: N'-(2,4-dimethylphenyl)-N-{[(2,4-dimethylphenyl)imino]methyl}-N-methylmethanimidamide
Synonyms: bipin 阿米曲士 triazid amitraz estrella mitaban fumilat A mitac mitaban (product) amitraz (substance) amitraz
Molecular Weight (g/mol): 293.41
Molecular Formula: C19H23N3
InChi Key: QXAITBQSYVNQDR-UHFFFAOYSA-N
SMILES: CN(C=NC1=CC=C(C)C=C1C)C=NC1=CC=C(C)C=C1C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results