accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOF

CAS RN 331-25-9

IUPAC Name: 2-(3-fluorophenyl)acetic acid
Synonyms: M-fluorophenylacetic acid
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: YEAUYVGUXSZCFI-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC(F)=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)