accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 331-62-4

IUPAC Name: 3-fluoro-4-methoxybenzonitrile
Synonyms: 3-fluoro-4-methoxybenzonitrile
Molecular Weight (g/mol): 151.14
Molecular Formula: C8H6FNO
InChi Key: FEEOVAOEPGQDTJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1F)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results