accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOH3COFFH3CH3CCH3CH3H3CCH3(E/Z)(E/Z)(E/Z)

CAS RN 331248-11-4

IUPAC Name: 7-[3,5-di-tert-butyl-2-(2,2-difluoroethoxy)phenyl]-3-methylocta-2,4,6-trienoic acid
Synonyms: 7-[3,5-di-tert-butyl-2-(2,2-difluoroethoxy)phenyl]-3-methylocta-2,4,6-trienoic acid
Molecular Weight (g/mol): 420.54
Molecular Formula: C25H34F2O3
InChi Key: BHIBZAZKKARFIM-UHFFFAOYSA-N
SMILES: CC(C=CC=C(C)C1=CC(=CC(=C1OCC(F)F)C(C)(C)C)C(C)(C)C)=CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results