accessibility menu, dialog, popup

UserName

Viewing profile as user:
MgBr

CAS RN 3315-91-1

IUPAC Name: {[1,1'-biphenyl]-4-yl}(bromo)magnesium
Synonyms: [1,1'-biphenyl]-4-yl(bromo)magnesium
Molecular Weight (g/mol): 257.41
Molecular Formula: C12H9BrMg
InChi Key: NACCJOIEZISYCU-UHFFFAOYSA-M
SMILES: Br[Mg]C1=CC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results