accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHN

CAS RN 33155-90-7

IUPAC Name: 1H,10H-cyclohexa[h]quinolin-10-one
Synonyms: 1H-cyclohexa[h]quinolin-10-one
Molecular Weight (g/mol): 195.22
Molecular Formula: C13H9NO
InChi Key: ZVFJWYZMQAEBMO-UHFFFAOYSA-N
SMILES: O=C1C=CC=C2C=CC3=CC=CNC3=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results