accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 331638-66-5

CH3NHOOH3C

CAS RN 331638-66-5

IUPAC Name: 4-methoxy-N-[(2-methylphenyl)methyl]benzamide
Synonyms: 4-methoxy-N-[(2-methylphenyl)methyl]benzamide
Molecular Weight (g/mol): 255.32
Molecular Formula: C16H17NO2
InChi Key: CVDCGDPYWBZBAN-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)NCC1=CC=CC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results