accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 331766-69-9

IUPAC Name: [(2-bromothiophen-3-yl)methyl](methyl)amine
Synonyms: [(2-bromothiophen-3-yl)methyl](methyl)amine
Molecular Weight (g/mol): 206.1
Molecular Formula: C6H8BrNS
InChi Key: FOIBDCULRFTGJL-UHFFFAOYSA-N
SMILES: CNCC1=C(Br)SC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results