accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONNNBr

CAS RN 331767-56-7

IUPAC Name: tert-butyl 4-(6-bromopyridin-2-yl)piperazine-1-carboxylate
Synonyms: tert-butyl 4-(6-bromopyridin-2-yl)piperazine-1-carboxylate
Molecular Weight (g/mol): 342.24
Molecular Formula: C14H20BrN3O2
InChi Key: CQSAPHUUCUUHNS-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=NC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results