accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONOOCl

CAS RN 331845-70-6

IUPAC Name: 2-chloro-N-cyclohexyl-N-methyl-4-nitrobenzamide
Synonyms: 2-chloro-N-cyclohexyl-N-methyl-4-nitrobenzamide
Molecular Weight (g/mol): 296.75
Molecular Formula: C14H17ClN2O3
InChi Key: FSSFCAMVJVCOMW-UHFFFAOYSA-N
SMILES: CN(C1CCCCC1)C(=O)C1=C(Cl)C=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results