accessibility menu, dialog, popup

UserName

Viewing profile as user:
SN

CAS RN 3319-99-1

IUPAC Name: 2-(thiophen-2-yl)pyridine
Synonyms: 2-(thiophen-2-yl)pyridine
Molecular Weight (g/mol): 161.22
Molecular Formula: C9H7NS
InChi Key: QLPKTAFPRRIFQX-UHFFFAOYSA-N
SMILES: S1C=CC=C1C1=CC=CC=N1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)