accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 331958-90-8

IUPAC Name: 4,4,5,5-tetramethyl-2-(oxolan-3-yl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(oxolan-3-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 198.07
Molecular Formula: C10H19BO3
InChi Key: KEBDNHHRQNZXMA-UHFFFAOYNA-N
SMILES: CC1(C)OB(OC1(C)C)C1CCOC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results