accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOBr

CAS RN 332-48-9

IUPAC Name: 1-(2-bromoethoxy)-4-fluorobenzene
Synonyms: 1-(2-bromoethoxy)-4-fluorobenzene
Molecular Weight (g/mol): 219.05
Molecular Formula: C8H8BrFO
InChi Key: JXSPKRUNMHMICQ-UHFFFAOYSA-N
SMILES: FC1=CC=C(OCCBr)C=C1
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)