accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOSF

CAS RN 332-51-4

IUPAC Name: 2-[(4-fluorophenyl)sulfanyl]acetate
Synonyms: 2-[(4-fluorophenyl)sulfanyl]acetate
Molecular Weight (g/mol): 185.19
Molecular Formula: C8H6FO2S
InChi Key: HQEROVXUKINLPI-UHFFFAOYSA-M
SMILES: [O-]C(=O)CSC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results