accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOOIOOOI

CAS RN 33239-19-9

IUPAC Name: disodium 4',5'-diiodo-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-3',6'-bis(olate)
Synonyms: disodium 4',5'-diiodo-3-oxospiro[2-benzofuran-1,9'-xanthene]-3',6'-bis(olate)
Molecular Weight (g/mol): 628.07
Molecular Formula: C20H8I2Na2O5
InChi Key: AHSJNHONMVUMLK-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C1=CC=C2C(OC3=C(I)C([O-])=CC=C3C22OC(=O)C3=CC=CC=C23)=C1I
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)