accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 332398-57-9

IUPAC Name: 4-(dihydroxyboranyl)naphthalene-1-carboxylic acid
Synonyms: 4-(dihydroxyboranyl)naphthalene-1-carboxylic acid
Molecular Weight (g/mol): 216
Molecular Formula: C11H9BO4
InChi Key: SRYYKYNBMCQCAB-UHFFFAOYSA-N
SMILES: OB(O)C1=C2C=CC=CC2=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results