accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33252-32-3

IUPAC Name: 2-amino-4-ethylpyridin-1-ium
Synonyms: 2-amino-4-ethylpyridin-1-ium
Molecular Weight (g/mol): 123.18
Molecular Formula: C7H11N2
InChi Key: SJWHILBZPGQBJE-UHFFFAOYSA-O
SMILES: CCC1=CC=[NH+]C(N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results